Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009074.hg.1 |
TC09000724.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009074.hg.1 |
ENST00000259339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009074.hg.1 |
ENST00000437263.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009074.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160564.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160564.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160564.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160564.hg.1 |
ENST00000259339.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160564.hg.1 |
ENST00000488169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160564.hg.1 |
NM_014506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160564.hg.1 |
TC0900008933.hg.1 |
Clariom D TC |