Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015092.4 | NP_055907.3 | 57 - 58 | 10436 | 71 | |
XM_005255182.3 | XP_005255239.1 | 56 - 57 | 10533 | 71 | ||
XM_005255183.3 | XP_005255240.1 | 57 - 58 | 10591 | 71 | ||
XM_005255184.3 | XP_005255241.1 | 55 - 56 | 10377 | 71 | ||
XM_011545769.2 | XP_011544071.1 | 56 - 57 | 10533 | 71 | ||
XM_011545770.1 | XP_011544072.1 | 44 - 45 | 8330 | 71 | ||
XM_017023066.1 | XP_016878555.1 | 56 - 57 | 10343 | 71 | ||
XM_017023067.1 | XP_016878556.1 | 44 - 45 | 8393 | 71 | ||
GenBank mRNA | AB007881.2 | - | 25 - 26 | 5054 | 71 | |
AB061371.1 | - | 58 - 59 | 10388 | 71 | ||
AF186377.1 | - | 32 - 33 | 6417 | 71 | ||
AF395444.1 | - | 55 - 56 | 9743 | 71 | ||
AY014957.1 | - | 45 - 46 | 8315 | 71 | ||
U32581.2 | - | 7 - 8 | 1446 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Homo sapiens Probe spans exons |