Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000470193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000470193.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000490419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
ENST00000490419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000225648.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000470193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000470193.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000484302.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000490419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
ENST00000490419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012433.hg.1 |
NM_018952.4 |
Refseq |