Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005499.hg.1 |
ENST00000215957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005499.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005499.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005499.hg.1 |
TC22000293.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000489812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000489812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000489812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
ENST00000489812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005500.hg.1 |
ENST00000215957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005500.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005500.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005500.hg.1 |
TC22000293.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
TC2200007324.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151542.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000445494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000489812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000489812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000489812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
ENST00000489812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007324.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
TC2200007324.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
NM_033386.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
ENST00000215957.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
ENST00000402631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
ENST00000424008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
ENST00000454685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151543.hg.1 |
NM_033386.3 |
Refseq |