Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022184.hg.1 |
TC09001731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022184.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022184.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022183.hg.1 |
TC09001731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022183.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022183.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182070.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182070.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182070.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182070.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182070.hg.1 |
TC0900011979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900182069.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182069.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182069.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182069.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182069.hg.1 |
TC0900011979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900182071.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182071.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182071.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182071.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182071.hg.1 |
TC0900011979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
ENST00000371712.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011979.hg.1 |
NM_019892.5 |
Refseq |