Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000463266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000463266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030902.hg.1 |
TC03001706.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030902.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030902.hg.1 |
ENST00000383659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030902.hg.1 |
ENST00000465044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030902.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030904.hg.1 |
TC03001706.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030904.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030904.hg.1 |
ENST00000383659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030904.hg.1 |
ENST00000465044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030904.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190888.hg.1 |
TC0300012195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000463266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000463266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000463266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000306103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000463266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000465044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000467643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
ENST00000478601.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
NM_024610.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190892.hg.1 |
TC0300012195.hg.1 |
Clariom D TC |