Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000328270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000328270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000328270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000328270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000328270.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399434.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399434.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399434.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399434.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000399537.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000413081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000413081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000413081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000413081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000413081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000430969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000434361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000434361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000434361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000434361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000440081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000440081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000440081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000440081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000455076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000455076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000455076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000455076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000455076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000464852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000464852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000464852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000464852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000497663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000497663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
ENST00000497663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010155.hg.1 |
NM_001080495.2 |
Refseq |