Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001014.hg.1 |
TC06000094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001014.hg.1 |
ENST00000451315.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001014.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001014.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147860.hg.1 |
TC0600006993.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000420088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000451315.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
ENST00000474485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006993.hg.1 |
NM_016167.4 |
Refseq |