Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001116.hg.1 |
TC06000108.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001116.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001116.hg.1 |
ENST00000356840.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001116.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
ENST00000584938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007060.hg.1 |
NR_039782.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148197.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148197.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148197.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148197.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148197.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148197.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148197.hg.1 |
TC0600007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000349606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000356840.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
ENST00000584938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NM_013262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007060.hg.1 |
NR_039782.2 |
Refseq |