Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008102.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007258.hg.1 |
TC06000606.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007258.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007258.hg.1 |
ENST00000451025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007258.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007258.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007258.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007257.hg.1 |
TC06000606.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007257.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007257.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007257.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156616.hg.1 |
TC0600008102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000451025.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_001003690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008102.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156615.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156615.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156615.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156615.hg.1 |
ENST00000372171.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156615.hg.1 |
ENST00000508232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156615.hg.1 |
NM_014628.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156615.hg.1 |
TC0600008102.hg.1 |
Clariom D TC |