Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026373.hg.1 |
ENST00000545018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026373.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026373.hg.1 |
TC11002204.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192557.hg.1 |
TC1100012012.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000279839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000539856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000542907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000616442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
ENST00000621358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012012.hg.1 |
NM_016403.3 |
Refseq |