Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000564667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000564667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000564667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
ENST00000564667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008760.hg.1 |
ENST00000507871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008760.hg.1 |
ENST00000507871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015279.hg.1 |
ENST00000303155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015279.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015279.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16015279.hg.1 |
TC16001088.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178477.hg.1 |
TC1600010181.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000564667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000564667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000564667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
ENST00000564667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010181.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008760.hg.1 |
ENST00000507871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008760.hg.1 |
ENST00000507871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
ENST00000303155.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
ENST00000562435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
ENST00000562559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
ENST00000563078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
NM_001201477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
NM_018092.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600178479.hg.1 |
TC1600010181.hg.1 |
Clariom D TC |