Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005025.hg.1 |
ENST00000307102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005025.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005025.hg.1 |
TC15000613.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000425818.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007614.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153987.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153987.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153987.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153987.hg.1 |
ENST00000307102.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153987.hg.1 |
ENST00000566326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153987.hg.1 |
NM_002755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153987.hg.1 |
TC1500007614.hg.1 |
Clariom D TC |