Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
ENST00000373005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_001008901.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_001008901.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_001008901.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_001008901.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_001008901.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_176791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_176791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_176791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_176791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_176791.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009169.hg.1 |
NM_176791.3 |
Refseq |