Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000490312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000490312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000490312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000490312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007383.hg.1 |
TC01000475.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007383.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007383.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007383.hg.1 |
ENST00000541606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007383.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100155182.hg.1 |
TC0100007838.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000296218.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000466723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000467277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000490312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000490312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000490312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000490312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
ENST00000497858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007838.hg.1 |
NM_003462.3 |
Refseq |