Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039111.hg.1 |
TC01002518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039111.hg.1 |
ENST00000296215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039111.hg.1 |
ENST00000436196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039111.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039111.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039112.hg.1 |
TC01002518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039112.hg.1 |
ENST00000296215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039112.hg.1 |
ENST00000436196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039112.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039112.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039116.hg.1 |
TC01002518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039116.hg.1 |
ENST00000296215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039116.hg.1 |
ENST00000436196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039116.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039116.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039114.hg.1 |
TC01002518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039114.hg.1 |
ENST00000296215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039114.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039117.hg.1 |
TC01002518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039117.hg.1 |
ENST00000296215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039117.hg.1 |
ENST00000436196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039117.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039113.hg.1 |
TC01002518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039113.hg.1 |
ENST00000296215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039113.hg.1 |
ENST00000436196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039113.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202509.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202509.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202509.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202509.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202509.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202509.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202509.hg.1 |
TC0100013747.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100202514.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202514.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202514.hg.1 |
TC0100013747.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100202512.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202512.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202512.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202512.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202512.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202512.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202512.hg.1 |
TC0100013747.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202508.hg.1 |
TC0100013747.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013747.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
ENST00000296215.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
ENST00000468040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
ENST00000493916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
NM_024700.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202510.hg.1 |
TC0100013747.hg.1 |
Clariom D TC |