Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026974.hg.1 |
ENST00000257570.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026974.hg.1 |
ENST00000339275.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026974.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026974.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026974.hg.1 |
TC12002059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000339275.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000543677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
ENST00000620239.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_001261825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012190.hg.1 |
NM_198213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195080.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195080.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195080.hg.1 |
ENST00000257570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195080.hg.1 |
NM_003733.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195080.hg.1 |
TC1200012190.hg.1 |
Clariom D TC |