Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005296.hg.1 |
TC09000491.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005296.hg.1 |
ENST00000259365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005296.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005296.hg.1 |
ENST00000395211.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005296.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005296.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154669.hg.1 |
TC0900008150.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000259365.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000375175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
ENST00000395211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_001166116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008150.hg.1 |
NM_003275.3 |
Refseq |