Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000469204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000469204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000469204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000479577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000479577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
ENST00000479577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000373189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000469204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000469204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000469204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000479577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000479577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
ENST00000479577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_001174098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NM_018344.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007954.hg.1 |
NR_033414.1 |
Refseq |