Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015129.hg.1 |
ENST00000374433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015129.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015129.hg.1 |
TC10001205.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015125.hg.1 |
ENST00000374433.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015125.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015125.hg.1 |
ENST00000475186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015125.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015125.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015125.hg.1 |
TC10001205.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007432.hg.1 |
ENST00000418966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007432.hg.1 |
ENST00000418966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007432.hg.1 |
ENST00000418966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007432.hg.1 |
ENST00000418966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007432.hg.1 |
NR_047558.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007432.hg.1 |
NR_047558.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007432.hg.1 |
NR_047558.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007432.hg.1 |
NR_047558.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171060.hg.1 |
TC1000010452.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000171063.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171063.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171063.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171063.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171063.hg.1 |
TC1000010452.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007432.hg.1 |
ENST00000418966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007432.hg.1 |
ENST00000418966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007432.hg.1 |
ENST00000418966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007432.hg.1 |
ENST00000418966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007432.hg.1 |
NR_047558.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007432.hg.1 |
NR_047558.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007432.hg.1 |
NR_047558.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007432.hg.1 |
NR_047558.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000374433.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000395797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
ENST00000485351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_001005368.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010452.hg.1 |
NM_006973.2 |
Refseq |