Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005793.hg.1 |
ENST00000264639.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005793.hg.1 |
ENST00000415039.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005793.hg.1 |
ENST00000540504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005793.hg.1 |
ENST00000541736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005793.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005793.hg.1 |
TC17000485.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000540504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
ENST00000580980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007772.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
ENST00000264639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
ENST00000415039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
ENST00000485835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
NM_002809.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700156390.hg.1 |
TC1700007772.hg.1 |
Clariom D TC |