Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005389.hg.1 |
TC09000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005389.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005389.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005391.hg.1 |
TC09000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005391.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005391.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005390.hg.1 |
TC09000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005390.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005390.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000470473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000470473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000470473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000615204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000615204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000615204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
ENST00000615204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000470473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000470473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000470473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000615204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000615204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000615204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
ENST00000615204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008197.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154889.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154889.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154889.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154889.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154889.hg.1 |
TC0900008197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900154890.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154890.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154890.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154890.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154890.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154890.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154890.hg.1 |
TC0900008197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900154891.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154891.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154891.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154891.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154891.hg.1 |
TC0900008197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900154892.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154892.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154892.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154892.hg.1 |
ENST00000375011.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154892.hg.1 |
ENST00000610463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154892.hg.1 |
NM_024642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154892.hg.1 |
TC0900008197.hg.1 |
Clariom D TC |