Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002266.hg.1 |
ENST00000254442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002266.hg.1 |
ENST00000357574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002266.hg.1 |
ENST00000398311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002266.hg.1 |
ENST00000444065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002266.hg.1 |
NM_015285.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002266.hg.1 |
NM_052834.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002266.hg.1 |
TC18000191.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
ENST00000254442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
ENST00000357574.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
ENST00000591524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
ENST00000615645.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
NM_015285.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
NM_052834.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
TC1800007385.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
ENST00000254442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
ENST00000357574.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
ENST00000591524.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
ENST00000615645.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
NM_015285.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150634.hg.1 |
NM_052834.2 |
Refseq |