Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018077.hg.1 |
TC0X001349.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018077.hg.1 |
ENST00000371126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018077.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018078.hg.1 |
TC0X001349.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018078.hg.1 |
ENST00000371126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018078.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018075.hg.1 |
TC0X001349.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018075.hg.1 |
ENST00000371126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018075.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010759.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010759.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010759.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010759.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010759.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010759.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173069.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173069.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173069.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173069.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173069.hg.1 |
TC0X00010759.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00173070.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173070.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173070.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173070.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173070.hg.1 |
TC0X00010759.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00173067.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173067.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173067.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173067.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173067.hg.1 |
TC0X00010759.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010759.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010759.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010759.hg.1 |
ENST00000371126.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010759.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010759.hg.1 |
NM_178470.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010759.hg.1 |
NM_178470.4 |
Refseq |