Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029589.hg.1 |
TC01001905.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029589.hg.1 |
ENST00000258243.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029589.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
ENST00000434387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011913.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187894.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187894.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187894.hg.1 |
ENST00000258243.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187894.hg.1 |
NM_014777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187894.hg.1 |
TC0100011913.hg.1 |
Clariom D TC |