Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004506.3 | NP_004497.1 | 11 - 12 | 1419 | 93 | |
GenBank mRNA | BC112323.1 | - | 11 - 12 | 1322 | 5197314 | 93 |
BC121051.1 | - | 11 - 12 | 1276 | 40117979 | 93 | |
M65217.1 | - | 11 - 12 | 1320 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |