Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006632.3 | NP_006623.2 | 3 - 4 | 414 | 72 | |
GenBank mRNA | AK296070.1 | - | 2 - 3 | 372 | 72 | |
AK298367.1 | - | 3 - 4 | 411 | 72 | ||
BC017952.1 | - | 3 - 4 | 4557795 | 72 | ||
U90545.1 | - | 3 - 4 | 623 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons |