Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020488.hg.1 |
TC10002931.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020488.hg.1 |
ENST00000370397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020488.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000585551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000585551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000585551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000585551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187787.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187787.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187787.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187787.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187787.hg.1 |
TC1000012585.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000370397.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000585551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000585551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000585551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000585551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000588656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
ENST00000590930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012585.hg.1 |
NM_001278.4 |
Refseq |