Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000494.3 | NP_000485.3 | 21 - 22 | 1936 | 108 | |
GenBank mRNA | AB900157.1 | - | 20 - 21 | 1767 | 108 | |
M63730.1 | - | 4 - 5 | 246 | 108 | ||
M91669.1 | - | 21 - 22 | 1872 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |