Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021420.hg.1 |
ENST00000353479.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021420.hg.1 |
ENST00000369733.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021420.hg.1 |
ENST00000541872.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021420.hg.1 |
NM_000494.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021420.hg.1 |
TC10001637.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000180350.hg.1 |
ENST00000353479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180350.hg.1 |
ENST00000369733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180350.hg.1 |
NM_000494.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180350.hg.1 |
ENST00000353479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180350.hg.1 |
ENST00000369733.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000180350.hg.1 |
NM_000494.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000180350.hg.1 |
TC1000011740.hg.1 |
Clariom D TC |