Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361803.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361803.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000361803.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000419159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000419159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
ENST00000419159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035398.hg.1 |
TC02002174.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035398.hg.1 |
ENST00000409382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035398.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035395.hg.1 |
TC02002174.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035395.hg.1 |
ENST00000361686.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035395.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035395.hg.1 |
ENST00000409382.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035395.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035395.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035395.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201028.hg.1 |
TC0200013779.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
ENST00000419159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
ENST00000419159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201029.hg.1 |
TC0200013779.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200201032.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201032.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201032.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201032.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201032.hg.1 |
TC0200013779.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361686.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361803.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361803.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000361803.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409087.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000409382.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000419159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000419159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
ENST00000419159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142351.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_001142352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013779.hg.1 |
NM_032528.2 |
Refseq |