Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145241.4 | NP_660284.1 | 3 - 4 | 418 | 94 | |
GenBank mRNA | BC012352.1 | - | 3 - 4 | 350 | 4634445 | 94 |
DA305733.1 | - | 3 - 4 | 341 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |