Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000562360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
ENST00000566795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011462.hg.1 |
NM_182563.3 |
Refseq |