Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF123654.1 | - | 2 - 3 | 561 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |