Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014669.hg.1 |
TC0X001074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014669.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X014669.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167037.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167037.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167037.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00167037.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00167037.hg.1 |
TC0X00009780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
ENST00000375135.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009780.hg.1 |
NM_004463.2 |
Refseq |