Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001590.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001590.hg.1 |
ENST00000540944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001590.hg.1 |
ENST00000558158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001590.hg.1 |
ENST00000559502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001590.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001590.hg.1 |
TC15000257.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
ENST00000560841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147732.hg.1 |
TC1500006890.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000319669.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558158.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000558625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000559813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
ENST00000560841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006890.hg.1 |
NM_152453.3 |
Refseq |