Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318245.1 | NP_001305174.1 | 22 - 23 | 4211 | 75 | |
NM_032608.6 | NP_115997.5 | 22 - 23 | 4208 | 75 | ||
XM_011530458.2 | XP_011528760.1 | 23 - 24 | 4347 | 75 | ||
XM_011530459.2 | XP_011528761.1 | 23 - 24 | 4318 | 75 | ||
XM_011530460.2 | XP_011528762.1 | 22 - 23 | 4191 | 75 | ||
XM_011530461.2 | XP_011528763.1 | 23 - 24 | 4347 | 75 | ||
XM_011530464.2 | XP_011528766.1 | 23 - 24 | 4344 | 75 | ||
XM_011530465.2 | XP_011528767.1 | 22 - 23 | 4104 | 75 | ||
XM_011530466.2 | XP_011528768.1 | 23 - 24 | 4348 | 75 | ||
XM_017029012.1 | XP_016884501.1 | 23 - 24 | 4347 | 75 | ||
XM_017029013.1 | XP_016884502.1 | 23 - 24 | 4395 | 75 | ||
XM_017029014.1 | XP_016884503.1 | 23 - 24 | 4347 | 75 | ||
XM_017029015.1 | XP_016884504.1 | 22 - 23 | 4218 | 75 | ||
XM_017029016.1 | XP_016884505.1 | 11 - 12 | 1471 | 75 | ||
XM_017029017.1 | XP_016884506.1 | 21 - 22 | 4072 | 75 | ||
GenBank mRNA | AB042648.2 | - | 18 - 19 | 2520 | 75 | |
AB075376.1 | - | 22 - 23 | 4208 | 75 | ||
AJ310931.2 | - | 22 - 23 | 4199 | 75 | ||
AY077700.1 | - | 22 - 23 | 4121 | 75 | ||
BC144596.1 | - | 20 - 21 | 3863 | 75 | ||
BC144597.1 | - | 21 - 22 | 4008 | 9053127 | 75 | |
BC150626.1 | - | 21 - 22 | 4008 | 9056996 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |