Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318245.1 | NP_001305174.1 | 25 - 26 | 4583 | 82 | |
NM_032608.6 | NP_115997.5 | 25 - 26 | 4580 | 82 | ||
XM_011530458.2 | XP_011528760.1 | 26 - 27 | 4719 | 82 | ||
XM_011530459.2 | XP_011528761.1 | 26 - 27 | 4690 | 82 | ||
XM_011530460.2 | XP_011528762.1 | 25 - 26 | 4563 | 82 | ||
XM_011530461.2 | XP_011528763.1 | 26 - 27 | 4719 | 82 | ||
XM_011530464.2 | XP_011528766.1 | 26 - 27 | 4716 | 82 | ||
XM_011530465.2 | XP_011528767.1 | 25 - 26 | 4476 | 82 | ||
XM_011530466.2 | XP_011528768.1 | 26 - 27 | 4720 | 82 | ||
XM_017029012.1 | XP_016884501.1 | 26 - 27 | 4719 | 82 | ||
XM_017029013.1 | XP_016884502.1 | 26 - 27 | 4767 | 82 | ||
XM_017029014.1 | XP_016884503.1 | 26 - 27 | 4719 | 82 | ||
XM_017029015.1 | XP_016884504.1 | 25 - 26 | 4590 | 82 | ||
XM_017029016.1 | XP_016884505.1 | 14 - 15 | 1843 | 82 | ||
XM_017029017.1 | XP_016884506.1 | 24 - 25 | 4444 | 82 | ||
GenBank mRNA | AB042648.2 | - | 21 - 22 | 2892 | 82 | |
AB075376.1 | - | 25 - 26 | 4580 | 82 | ||
AJ310931.2 | - | 25 - 26 | 4571 | 82 | ||
AY077700.1 | - | 25 - 26 | 4493 | 82 | ||
BC144596.1 | - | 23 - 24 | 4235 | 82 | ||
BC144597.1 | - | 24 - 25 | 4380 | 9053127 | 82 | |
BC150626.1 | - | 24 - 25 | 4380 | 9056996 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |