Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318245.1 | NP_001305174.1 | 37 - 38 | 6097 | 61 | |
NM_032608.6 | NP_115997.5 | 37 - 38 | 6094 | 61 | ||
XM_011530458.2 | XP_011528760.1 | 38 - 39 | 6233 | 61 | ||
XM_011530459.2 | XP_011528761.1 | 38 - 39 | 6204 | 61 | ||
XM_011530460.2 | XP_011528762.1 | 37 - 38 | 6077 | 61 | ||
XM_011530461.2 | XP_011528763.1 | 38 - 39 | 6233 | 61 | ||
XM_011530464.2 | XP_011528766.1 | 38 - 39 | 6230 | 61 | ||
XM_011530465.2 | XP_011528767.1 | 37 - 38 | 5990 | 61 | ||
XM_011530466.2 | XP_011528768.1 | 38 - 39 | 6234 | 61 | ||
XM_017029012.1 | XP_016884501.1 | 38 - 39 | 6233 | 61 | ||
XM_017029013.1 | XP_016884502.1 | 38 - 39 | 6281 | 61 | ||
XM_017029014.1 | XP_016884503.1 | 38 - 39 | 6230 | 61 | ||
XM_017029015.1 | XP_016884504.1 | 37 - 38 | 6101 | 61 | ||
XM_017029016.1 | XP_016884505.1 | 26 - 27 | 3357 | 61 | ||
XM_017029017.1 | XP_016884506.1 | 36 - 37 | 5958 | 61 | ||
GenBank mRNA | AB042648.2 | - | 33 - 34 | 4406 | 61 | |
AB075376.1 | - | 37 - 38 | 6094 | 61 | ||
AJ310931.2 | - | 37 - 38 | 6085 | 61 | ||
AY077700.1 | - | 37 - 38 | 6007 | 61 | ||
BC144596.1 | - | 35 - 36 | 5749 | 61 | ||
BC144597.1 | - | 36 - 37 | 5891 | 9053127 | 61 | |
BC150626.1 | - | 36 - 37 | 5891 | 9056996 | 61 | |
CR749556.1 | - | 2 - 3 | 140 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |