Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013210.hg.1 |
TC09001008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013210.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013210.hg.1 |
ENST00000541701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013210.hg.1 |
ENST00000541851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013210.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167365.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167365.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167365.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167365.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167365.hg.1 |
TC0900009846.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000379731.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
ENST00000535206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009846.hg.1 |
NM_001497.3 |
Refseq |