Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005315.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005315.hg.1 |
ENST00000400018.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005315.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005315.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005315.hg.1 |
TC13000468.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000475251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000475251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000475251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000475251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000536239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000536239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000536239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
ENST00000536239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000462482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000465471.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000475251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000475251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000475251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000475251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000536239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000536239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000536239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
ENST00000536239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008280.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
ENST00000298260.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
ENST00000314759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
ENST00000400018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
NM_001166017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
NM_145061.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300155888.hg.1 |
TC1300008280.hg.1 |
Clariom D TC |