Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032888.3 | NP_116277.2 | 14 - 15 | 2872 | 97 | |
XM_006717308.2 | XP_006717371.1 | 13 - 14 | 2886 | 97 | ||
XM_006717310.3 | XP_006717373.1 | 12 - 13 | 685 | 97 | ||
XM_011519138.2 | XP_011517440.1 | 14 - 15 | 2504 | 97 | ||
XM_011519140.1 | XP_011517442.1 | 14 - 15 | 2940 | 97 | ||
XM_011519142.2 | XP_011517444.1 | 14 - 15 | 2946 | 97 | ||
XM_011519143.2 | XP_011517445.1 | 14 - 15 | 2944 | 97 | ||
XM_011519144.2 | XP_011517446.1 | 14 - 15 | 2944 | 97 | ||
XM_017015239.1 | XP_016870728.1 | 14 - 15 | 2945 | 97 | ||
GenBank mRNA | AB058773.1 | - | 11 - 12 | 1360 | 97 | |
AY149237.1 | - | 14 - 15 | 2466 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |