Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCHY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278539.1 | NP_001265468.1 | 3 - 4 | 511 | 133 | |
NR_037914.1 | 3 - 4 | 469 | 133 | |||
NR_103725.1 | 2 - 3 | 247 | 133 | |||
XM_011531838.2 | XP_011530140.1 | 3 - 4 | 442 | 133 | ||
XM_011531839.2 | XP_011530141.1 | 3 - 4 | 630 | 133 | ||
GenBank mRNA | AK309555.1 | - | 3 - 4 | 316 | 133 | |
BF102905.1 | - | 2 - 3 | 247 | 133 | ||
BG703058.1 | - | 3 - 4 | 342 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |