Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206846.1 | NP_001193775.1 | 23 - 24 | 5329 | 86 | |
NM_018136.4 | NP_060606.3 | 24 - 25 | 10084 | 86 | ||
GenBank mRNA | AF509326.1 | - | 24 - 25 | 9827 | 86 | |
AK001379.1 | - | 7 - 8 | 3218 | 86 | ||
AK001411.1 | - | 7 - 8 | 1611 | 86 | ||
AK226178.1 | - | 23 - 24 | 5261 | 86 | ||
AY367065.1 | - | 24 - 25 | 9827 | 86 | ||
AY971955.1 | - | 10 - 11 | 2582 | 86 | ||
AY971956.1 | - | 23 - 24 | 5072 | 86 | ||
AY971957.1 | - | 10 - 11 | 3785 | 86 | ||
BC034607.1 | - | 19 - 20 | 2948 | 86 | ||
BC040439.1 | - | 7 - 8 | 1646 | 86 | ||
BX648804.1 | - | 7 - 8 | 4938 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |