Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000346.hg.1 |
TC0X000025.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000346.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000346.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000345.hg.1 |
TC0X000025.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000345.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000345.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000347.hg.1 |
TC0X000025.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000347.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000347.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177187.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177187.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177187.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177187.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177187.hg.1 |
TC0X00011259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00177185.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177185.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177185.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177185.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177185.hg.1 |
TC0X00011259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006657.hg.1 |
ENST00000412493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177186.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177186.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177186.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177186.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177186.hg.1 |
TC0X00011259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |