Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007027.3 | NP_008958.2 | 26 - 27 | 4400 | 150 | |
XM_005247076.3 | XP_005247133.1 | 26 - 27 | 4872 | 150 | ||
XM_017005636.1 | XP_016861125.1 | 26 - 27 | 4470 | 150 | ||
XM_017005637.1 | XP_016861126.1 | 23 - 24 | 3924 | 150 | ||
GenBank mRNA | AB019397.1 | - | 26 - 27 | 4347 | 150 | |
AK026229.1 | - | 4 - 5 | 501 | 150 | ||
AK302584.1 | - | 26 - 27 | 4366 | 150 | ||
BC126209.1 | - | 25 - 26 | 4290 | 8991925 | 150 | |
BC151237.1 | - | 26 - 27 | 4353 | 8860504 | 150 | |
D87448.1 | - | 26 - 27 | 4353 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |