Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037706.hg.1 |
TC01002450.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037706.hg.1 |
ENST00000263694.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037706.hg.1 |
ENST00000446633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037706.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037706.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000463988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000463988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000463988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000491106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000491106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
ENST00000491106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100200647.hg.1 |
TC0100013557.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000263694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000373720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000463988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000463988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000463988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000474025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000486941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000489853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000491106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000491106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
ENST00000491106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013557.hg.1 |
NM_004814.2 |
Refseq |