Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182237.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182237.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182237.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182237.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182237.hg.1 |
TC1400010737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000355081.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000396128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000556500.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000557468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
ENST00000579157.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_001308112.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010737.hg.1 |
NM_017658.4 |
Refseq |