Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033026.1 | NP_001028198.1 | 10 - 11 | 1462 | 89 | |
XM_005259678.2 | XP_005259735.1 | 10 - 11 | 1507 | 89 | ||
XM_017027457.1 | XP_016882946.1 | 10 - 11 | 1456 | 89 | ||
GenBank mRNA | AK090400.1 | - | 6 - 7 | 527 | 89 | |
AY140906.1 | - | 1 - 2 | 102 | 89 | ||
DQ005958.1 | - | 10 - 11 | 1444 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens 1700 Probe spans exons |