Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011834.hg.1 |
TC01000736.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011834.hg.1 |
ENST00000282670.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011834.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000448074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000448074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000448074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000448074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000489510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008637.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
ENST00000282670.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
ENST00000491611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
ENST00000525663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
ENST00000528352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
NM_152665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161442.hg.1 |
TC0100008637.hg.1 |
Clariom D TC |